Learn More
Medchemexpress LLC Saxagliptin hydrochloride | 709031-78-7 | 99.7% | 351.87 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16448100MG
Encompass_Preferred
Saxagliptin hydrochloride (BMS-477118 hydrochloride) is a potent, selective, reversible, competitive and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor with a Ki of 0.6-1.3 nM. It has the potential for type 2 diabetes mellitus research.
  • Potent, selective, reversible, competitive, and orally active dipeptidyl peptidase-4 (DPP-4) inhibitor.
  • Potential for type 2 diabetes mellitus research.
Catalog No. 50-003-92353