Learn More
Medchemexpress LLC Saxagliptin hydrochloride | 709031-78-7 | MFCD22572334 | 99.7% | 351.87 g/mol | C18H26ClN3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1644810MG
Encompass_Preferred
Saxagliptin hydrochloride is the hydrochloride salt of saxagliptin, a potent and selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied as a high-purity solid for laboratory research.
  • High purity suitable for research applications.
  • Potent DPP-4 inhibitor for enzymatic and cellular assays.
  • Supplied in small mg quantities for compound screening and early-stage studies.
  • Well-characterized chemical identity (CAS: 709031-78-7).
  • Molecular formula C18H26ClN3O2; molecular weight 351.87 g/mol.
Catalog No. 50-002-89111