Learn More
Sigma Aldrich Fine Chemicals Biosciences SB 12 L Shaking Water Bath1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7414151EA
Encompass
This compact shaking water bath utilizes the well proven belt-less motor drive system from our popular Orbi-Shaker series with the addition of precision temperature control up to 80C.The seamless stainless steel chamber has a 12 L capacity and includes a universal spring-loaded platform for securing 15 50 mL test tubes as well as Erlenmeyer flasks and media bottles up to 1 L.A hinged and removable lid is also included to reduce energy waste prevent splashing and divert condensation away from samples.Easy to use controls allow the user to digitally select and monitor time temperature and shaking speed while a constant monitoring system ensures accuracy over each parameter.Included Spring platform accepts tubes bottles and flasks
Catalog No. 50-290-8885