Learn More
Enzo Life Sciences SB-202190 (1mg). CAS: 152121-30-7
SDP
Supplier:  Enzo Life Sciences BMLEI2940001
Encompass_Preferred
Cell permeable, potent and selective p38 MAP kinase inhibitor (IC50=30 nM). Inhibits p38α and ß, but not γ and δ isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. A useful tool for dissecting the role of p38 in signaling pathways. Induces apoptosis. Alternative name: FHPI, 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)-1H-imidazole. Purity: ≥98% (TLC). Solubility: Soluble in methanol, DMSO or acetone; slightly soluble in ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-201-0676