Learn More
TARGETMOL CHEMICALS INC SB 202190 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2301200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SB 202190 (FHPI) is a p38 MAPK inhibitor that inhibits p38alpha and p38beta2 (IC50=50/100 nM) selectively and cell-permeably. SB 202190 has antitumor activity and also induces the differentiation of human embryonic stem cells into cardiomyocytes. Purity 99.67%
Catalog No. 50-287-7570