Learn More
Medchemexpress LLC SB 202190 hydrochloride | 350228-36-3 | MFCD17215958 | 99.7% | 367.80 g/mol | C20H15ClFN3O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10295A5MG
Encompass_Preferred
SB 202190 hydrochloride is the hydrochloride salt of a selective p38 mitogen-activated protein kinase (MAPK) inhibitor used in biochemical and cellular research to probe p38-dependent signaling. It is supplied as a high-purity solid for in vitro and preclinical studies.
  • Selective p38 MAPK inhibitor for cell signaling and biochemical assays.
  • High purity (99.66%) suitable for analytical and biological experiments.
  • Solid form, easily dissolved in DMSO for stock solutions.
  • Available in small research quantities (e.g., 5 mg) and larger pack sizes.
  • Includes supporting analytical data such as COA, HNMR, and LCMS.
Catalog No. 50-002-48986