Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB-203186 hydrochloride | 207572-69-8 | MFCD04037547 | 99.9% | 308.80 g/mol | C16H21ClN2O2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY10122210MM 1ML

Encompass_Preferred

SB-203186 hydrochloride is a potent, selective, competitive antagonist of the 5-HT4 serotonin receptor used in pharmacology research to probe receptor-mediated responses in tissue and cell-based assays. It is provided as a solid or as a pre-made solution for convenient in vitro use.

  • Potent, selective 5-HT4 receptor antagonist.
  • High purity (99.9%).
  • Molecular weight 308.80 g/mol.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solids in multiple sizes.
  • Soluble in DMSO up to 100 mg/mL; may require ultrasonic treatment.
  • Store sealed, away from moisture and light; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-12767


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.