Learn More
Apexbio Technology LLC SB 203580 hydrochloride, 869185-85-3, MFCD01862622, 25 mg
SDP
Supplier:  Apexbio Technology LLC B128525
Encompass_Preferred
MF: C21H17ClFN3OS, MW: 413.9, Solubility: 20.7mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. SB203580 HCl is a specific inhibitor of p38-MAPKs with IC50 value of 0.6 uM.
Catalog No. 50-101-2181