Learn More
Medchemexpress LLC SB 206553 hydrochloride | 1197334-04-5 | 99.4% | 328.80 g·mol⁻1 | C17H17ClN4O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1031355MG
Encompass_Preferred
SB 206553 hydrochloride is the hydrochloride salt of SB 206553, a selective, orally active antagonist of serotonin 5-HT2B and 5-HT2C receptors used in preclinical receptor pharmacology and behavioral studies.
  • High-affinity 5-HT2B/5-HT2C receptor antagonist.
  • Hydrochloride salt form for improved solubility.
  • High purity suitable for research (99.4%).
  • Available in small research quantities, such as 5 mg.
Catalog No. 50-002-45788