Learn More
Enzo Life Sciences SB-216763 (1mg). CAS: 280744-09-4
SDP
Supplier:  Enzo Life Sciences BMLEI3120001
Encompass_Preferred
Potent and selective inhibitor of GSK-3β (IC50=34nM). Stimulates glycogen synthesis in human liver cells and mimics other actions of insulin. Protects primary neurons from death induced by PI-3 kinase pathway inhibition suggesting therapeutic potential in neurodegenerative diseases and stroke. Reduces pulmonary inflammation and fibrosis in a mouse model. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0700