Learn More
Medchemexpress LLC HY-12012 5mg Medchemexpress, SB 216763 CAS:280744-09-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120125MG
Encompass_Preferred
Medchemexpress, HY-12012 5mg SB 216763 CAS:280744-09-4 SB 216763 is potent, selective and ATP-competitive GSK-3 inhibitor with IC50s of 34.3 nM for both GSK-3α and GSK-3β. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9581