Learn More
Medchemexpress LLC SB-224289 hydrochloride | 180084-26-8 | 99.0% | 557.08 g/mol | C32H33ClN4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101105A5MG
Encompass_Preferred
A selective 5-HT1B receptor antagonist used in pharmacological research to study serotonin receptor function and behavioral effects. The hydrochloride salt is supplied as a white to off-white solid with high purity and documented solubility and storage conditions for in vitro and in vivo applications.
  • Selective 5-HT1B receptor antagonist with reported anxiolytic effects.
  • High affinity for 5-HT1B receptors (pKi ≈ 8).
  • More than 80-fold selectivity over related receptors, improving assay specificity.
  • Suitable for in vitro and in vivo studies, with recommended formulation guidance.
  • High purity (99.0%), minimizing confounding impurities.
  • Soluble in DMSO and formulated for several in vivo vehicles.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-002-13517