Learn More
Cayman Chemical SB-224289hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1703610 MG
Encompass_Preferred
A potent 5-HT1B inverse agonist (pKi = 8.16) that displays >75-fold selectivity for human 5-HT1B over all other 5-HT receptors
Catalog No. 50-296-6513