Learn More
Abcam SB 225002, non-peptide C x CR2 antagonist, 10MG
Supplier:  Abcam AB12089510MG
Encompass_Preferred
MW 352.14 g/mol, Purity >95%. A selective non-peptide CXCR2 antagonist. Inhibits IL-8 binding to CXCR2 (IC₅₀ = 22 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7157