Learn More
Medchemexpress LLC SB 239063 | 193551-21-2 | 99.2% | 368.40 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1106850MG
Encompass_Preferred
SB 239063 is a potent, selective and orally active p38 MAPK inhibitor. It exhibits an IC50 of 44 nM for recombinant purified human p38α, with equipotent inhibitory activity against p38α and p38β. SB 239063 has no effect on p38γ or p38δ.
  • Potent, selective, and orally active p38 MAPK inhibitor.
  • Exhibits equipotent inhibitory activity against p38α and p38β.
  • Has anti-asthma activity.
  • Can enhance memory impaired due to aging or medical conditions, such as AD.
  • Able to penetrate the blood brain barrier (BBB).
Catalog No. 50-003-43232