Learn More
Abcam SB 242084 hydrochloride, 5-HT2C antagonist, 10MG
Supplier:  Abcam AB12051910MG
Encompass_Preferred
MW 431.3 Da, Purity >98%. Potent, selective 5-HT2C antagonist (pKi values are 9 (5-HT2C), 7 (5-HT2B), 6.8 (5-HT2A), 6.4 (5-HT1A), 6.4 (5-HT1B), 6.4 (5-HT1D), 6 (5-HT1E), <6.1 (5-HT1F), <5.5 (5-HT4), 6 (5-HT6), 6.1 (5-HT7), 6.2 (D2), 6.2 (D3) and <5 (α1)). Blood-brain barrier permeable. Anxiolytic activity *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4588