Learn More
Selleck Chemical LLC SB 265610 S9516-25MG
SDP
Supplier:  Selleck Chemical LLC S951625MG
Encompass_Preferred
SB265610 a competitive antagonist at the human CXCR2 receptor can displace [125I]-IL-8 and [125I]-GRO with pIC50 values of 8 41 and 8 47 respectively preventing receptor activation by binding to a region distinct from the agonist binding site
Catalog No. 50-000-23992