Learn More
Medchemexpress LLC SB-269970 hydrochloride | 261901-57-9 | C18H29ClN2O3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15370A50MG
Encompass_Preferred
SB-269970 hydrochloride is a potent, selective, and brain-penetrant 5-HT7 receptor antagonist. It exhibits >50-fold selectivity against other 5-HT receptors, making it suitable for research into serotonin pathways and neurological conditions.
  • Potent and selective 5-HT7 receptor antagonist
  • Brain-penetrant
  • High selectivity against other 5-HT receptors
  • High purity
  • White to off-white solid
  • Molecular weight 388.95
Catalog No. 50-003-76149