Learn More
Abcam SB 269970 hydrochloride, 5-HT7 receptor antagonist, 10MG
Supplier:  Abcam AB12050810MG
Encompass_Preferred
MW 389 Da, Purity >98%. Potent, selective 5-HT7 receptor antagonist (pKi values are 8.3, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B receptors, respectively). Also potently blocks α2 adrenoceptors. Blocks amphetamine and ketamine induced hyperactivity. Brain penetrant *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7537