Learn More
Abcam SB 271046 hydrochloride, 5-HT6 antagonist, 50MG
Supplier:  Abcam AB12058950MG
Encompass_Preferred
MW 488.45 g/mol, Purity >99%. Potent and selective 5-HT₆ receptor antagonist (Ki = 1-2 nM). Displays >200-fold selectivity for 5-HT₆ receptors (vs. other receptor binding sites and ion channels). Poor penetration of the blood-brain barrier. Nootropic effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7455