Learn More
Cayman Chemical SB-271046hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1851310 MG
Encompass_Preferred
A selective, orally-available antagonist of the 5-HT6 receptor (pKi = 9.02-8.92); produces a significant increase in extracellular levels of glutamate and aspartate within the frontal cortex; used to evaluate the role of 5-HT6 in learning and memory
Catalog No. 50-296-6008