Learn More
Cayman Chemical SB 277011AhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 119825 MG
Encompass_Preferred
A selective antagonist of the dopamine D3 receptor (pKi = 8.0); has high oral bioavailability and enters the central nervous system of the rat
Catalog No. 50-295-4924