Learn More
Abcam SB-3CT, MMP-2 and MMP-9 inhibitor, 1MG
Supplier:  Abcam AB1415791MG
Encompass_Preferred
MW 306.4 Da, Purity >98%. Potent, competitive and non-selective MMP-2 and MMP-9 inhibitor (Ki values are 13.9 and 600 nM for MMP-2 and MMP-9 respectively). Does not alter the activity of MMP-1, MMP-3 or MMP-7 (Ki values are 206 (MMP-1), 15 (MMP-3) and 96 (MMP-7) μM). Neuroprotective and blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6512