Learn More
Enzo Life Sciences SB-415286 (5mg). CAS: 264218-23-7
SDP
Supplier:  Enzo Life Sciences BMLEI3110005
Encompass_Preferred
Potent and selective inhibitor of GSK-3β (IC50=78nM). Stimulates glycogen synthesis in human liver cells and mimics other actions of insulin. Protects primary neurons from death induced by PI-3 kinase pathway inhibition suggesting therapeutic potential in neurodegenerative diseases and stroke. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0699