Learn More
Medchemexpress LLC SB-423562 | 351490-27-2 | C26H32N2O4 | 10 MM * 1 ML

Supplier: Medchemexpress LLC HY1510510MM/1ML
SB-423562 is a short-acting calcium-sensing receptor (CaR) antagonist. It has potential for osteoporosis research.
- Short-acting calcium-sensing receptor (CaR) antagonist
- Supports osteoporosis research
- Purity of 99.66%
- Supplied as 10 MM solution in 1 ML of DMSO
- Powder storage: -20°C for 3 years; 4°C for 2 years
- Solution storage: -80°C for 2 years; -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.