Learn More
Cayman Chemical SB-431542, 301836-41-9, 10 mg
SDP
Supplier:  Cayman Chemical 1303110
Encompass_Preferred
Molecular Formula C22H16N4O3, Purity ≥98%, Formula Weight 384.4; A potent and selective inhibitor of the TGF-1 receptor ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM) and, less effectively, ALK7; suppresses renewal in embryonic and induced pluripotent stem cells and promotes their differentiation.
Catalog No. NC1390838