Learn More
Selleck Chemical LLC SB431542 10mg 301836-41-9
SDP
Supplier:  Selleck Chemical LLC S106710MG
Encompass_Preferred
SB431542 is a potent and selective inhibitor of ALK5 with IC50 of 94 nM in a cell-free assay, 100-fold more selective for ALK5 than p38 MAPK and other kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-797-0