Learn More
Cayman Chemical SB-431542hydrate 10mg
SDP
Supplier:  Cayman Chemical 1303110 MG
Encompass_Preferred
A potent and selective inhibitor of the TGF-β1 receptor ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM) and, less effectively, ALK7; suppresses renewal in embryonic and induced pluripotent stem cells and promotes their differentiation
Catalog No. 50-295-3255