Learn More
TARGETMOL CHEMICALS INC SB 525334 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T176325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-525334 is a potent and selective inhibitor of the TGF-(beta)1R and ALK5 (IC50 14.3 nM). purity: 99%
Catalog No. 50-258-1099