Learn More
Medchemexpress LLC Sb 525334 | 356559-20-1 | 100.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY12043500MG
Encompass_Preferred
A potent, selective inhibitor of the transforming growth factor β type I receptor (ALK5) with nanomolar biochemical potency. Supplied as high-purity solid and DMSO solution formats for research use; consult the SDS for handling and safety guidance.
  • Potent and selective ALK5 inhibitor with IC50 of 14.3 nM.
  • High purity suitable for research applications.
  • Available as solid and 10 mM DMSO solution formats, including 500 mg solid packs.
  • Molecular weight 343.42 g/mol and formula C21H21N5.
  • Stable when stored at recommended temperatures for long-term storage.
  • SDS available with handling, personal protective equipment, and first-aid instructions.
Catalog No. 50-002-34165