Learn More
Medchemexpress LLC SB-612111 hydrochloride | 371980-94-8 | 99.6% | 454.86 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18618A
Encompass_Preferred
SB-612111 hydrochloride is a novel and potent opiate receptor-like orphan receptor (ORL-1) antagonist. It demonstrates high affinity for hORL-1 and selectivity for μ-, κ- and δ-receptors. This compound effectively antagonizes the pronociceptive action of Nociceptin in acute pain models.
  • Potent opiate receptor-like orphan receptor (ORL-1) antagonist
  • High affinity for hORL-1 (Ki=0.33 nM)
  • Exhibits selectivity for μ-, κ- and δ-receptors
  • Antagonizes pronociceptive action of Nociceptin
  • Had antihyperalgesic effects on carrageenan-induced rat paw
Catalog No. 50-003-84149