Learn More
Medchemexpress LLC HY-12069 5mg Medchemexpress, SB-743921 CAS:940929-33-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120695MG
Encompass_Preferred
Medchemexpress, HY-12069 5mg SB-743921 CAS:940929-33-9 SB-743921 is a potent inhibitor of the mitotic kinesin KSP (Eg5), with a Ki of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9890