Learn More
Selleck Chemical LLC SB743921 HCl 50mg 940929-33-9
SDP
Supplier:  Selleck Chemical LLC S218250MG
Encompass_Preferred
SB743921 is a kinesin spindle protein (KSP) inhibitor with Ki of 0.1 nM, almost no affinity to MKLP1, Kin2, Kif1A, Kif15, KHC, Kif4 and CENP-E. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-859-6