Learn More
TARGETMOL CHEMICALS INC SB-747651A DIHYDROCHLORI 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T96525MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SB-747651A dihydrochloride is an ATP-competitive mitogen- and stress-activated kinase 1 (MSK1) inhibitor with an IC 50 of 11 nM. SB-747651A dihydrochloride also inhibits PRK2 RSK1 p70S6K and ROCK-II. SB-747651A dihydrochloride has potential for inflammation research [1]. purity: 97%
Catalog No. 50-257-8981