Learn More
Medchemexpress LLC SB-747651A (dihydrochloride) | 1781882-72-1 | 99.14% | 415.32 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY110313
Encompass_Preferred
SB-747651A dihydrochloride is an ATP-competitive inhibitor of mitogen- and stress-activated kinase 1 (MSK1) with an IC₅₀ of 11 nM. It also inhibits PRK2, RSK1, p70S6K, and ROCK-II. This compound is used in inflammation research.
  • ATP-competitive MSK1 inhibitor
  • Inhibits PRK2, RSK1, p70S6K, and ROCK-II
  • Suitable for inflammation research
  • Modulates chemokine-induced neutrophil recruitment
  • Affects neutrophil extravasation in acute inflammation models
Catalog No. 50-003-45763