Learn More
Medchemexpress LLC SB-747651A dihydrochloride | 1781882-72-1 | MFCD26792571 | 99.1% | 415.32 g/mol | C16H24Cl2N8O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1103135MG
Encompass_Preferred
SB-747651A dihydrochloride is an ATP-competitive inhibitor of mitogen- and stress-activated kinase 1 (MSK1) with potent in vitro activity. Supplied as the dihydrochloride salt, it is intended for kinase profiling and inflammation-related research applications.
  • ATP-competitive MSK1 inhibitor with IC50 11 nM.
  • Also active against PRK2, RSK1, p70S6K, and ROCK-II.
  • Dihydrochloride salt form, solid white to off-white.
  • High purity (99.1%) suitable for biochemical assays.
  • Soluble in water approximately 50 mg/mL; may require ultrasonic.
  • Store sealed at 4°C; in solvent store -80°C up to 6 months.
  • Available in small research pack sizes such as 5 mg.
Catalog No. 50-002-63598