Learn More
Medchemexpress LLC SB-772077B dihydrochloride | 607373-46-6 | 99.3% | 415.28 g/mol | C15H20Cl2N8O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1085185MG
Encompass_Preferred
Aminofuran-based dihydrochloride Rho-associated kinase (ROCK) inhibitor for research use. It shows low-nanomolar potency against ROCK1 and ROCK2 and is used in studies of vascular tone, inflammatory signaling, and blood pressure regulation. The compound is supplied at high purity and is appropriate for biochemical and cellular assays.
Catalog No. 50-002-63691