Learn More
Perkin Elmer US LLC Antimony (Sb) Lumina Hollow Cathode Lamp with Intensitron Cable
Supplier:  Perkin Elmer US LLC N3050370
Encompass_Preferred
Single Element Lumina hollow cathode lamp (HCL) with Intensitron adapter cable for the detection of elemental Antimony (Sb).Please note that this lamp requires the use of a nitrous oxide-acetylene flame. Every genuine PerkinElmer lamp is designed for use with and tested on PerkinElmer spectrometers to assure compatibility and the highest performance. PerkinElmer offers a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume — and the longer the lamp lifetime.
Catalog No. 50-904-4334