Learn More
AdipoGen SB202190
SDP
Supplier:  AdipoGen AGCR10028M005
Encompass_Preferred
Chemical. CAS 152121-30-7. Formula C20H14FN3O. MW 331.3. Potent and cell permeable p38 MAP kinase inhibitor. Apoptosis inducer. Inhibits p38alpha and beta, but not gamma and delta isoforms. Does not inhibit ERK2 or other members of the MAP kinase family or their upstream activators. JNK activator. Autophagic vacuole inducer
Catalog No. 50-256-2388