Learn More
Selleck Chemical LLC SB202190 (FHPI) 10mM/1mL 152121-30-7
SDP
Supplier:  Selleck Chemical LLC S107710MM/1ML
Encompass_Preferred
SB202190 (FHPI) is a potent p38 MAPK inhibitor targeting p38?/? with IC50 of 50 nM/100 nM in cell-free assays, sometimes used instead of SB 203580 to investigate potential roles for SAPK2a/p38 in vivo. SB202190 inhibits endothelial cell apoptosis via induction of autophagy and heme oxygenase-1. SB202190 significantly suppresses Erastin?dependent ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0897