Learn More
Enzo Life Sciences SB202190. hydrochloride (5mg). CAS: 350228-36-3
SDP
Supplier:  Enzo Life Sciences ALX270268M005
Encompass_Preferred
A water soluble form of the potent p38 (SAPK2a) inhibitor SB202190 (BML-EI294). Alternative name: FHPI . HCl, 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)-1H-imidazole . HCl. Formula: C20H14FN3O . HCl. MW: 331.3 . 36.5. Purity: ≥96% (HPLC). Appearance: Yellow solid. Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8455