Learn More
Selleck Chemical LLC SB239063 5mg 193551-21-2
SDP
Supplier:  Selleck Chemical LLC S77415MG
Encompass_Preferred
SB239063 is a potent and selective p38 MAPK?/? inhibitor with IC50 of 44 nM, showing no activity against the ?- and ?-kinase isoforms. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5196