Learn More
Selleck Chemical LLC SB269970 HCl 10mg 261901-57-9
SDP
Supplier:  Selleck Chemical LLC S284910MG
Encompass_Preferred
SB269970 HCl is a hydrochloride salt form of SB-269970, which is a 5-HT7 receptor antagonist with pKi of 8.3, exhibits >50-fold selectivity against other receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3056