Learn More
Selleck Chemical LLC SB290157 trifluoroacetate 5mg
SDP
Supplier:  Selleck Chemical LLC S89315MG
Encompass_Preferred
SB290157 trifluoroacetate (SB 290157 TFA) is a potent, competitive and selective antagonist of C3a receptor (C3aR) with IC50 of 200 nM for RBL-C3aR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3441