Learn More
Apexbio Technology LLC SB408124, 100mg Cas# 288150-92-5
SDP
Supplier:  Apexbio Technology LLC B2163100
Encompass_Preferred
SB408124 is a potent and novel nonpeptide antagonist of orexin-1 (OX(1)) receptor with Ki values of 27 nM and 57 nM in membrane formats and in whole cell, respectively. SB408124 was 50-fold selective over the human OX2 receptor. 288150-92-5. MFCD06411575 SB408124 is a potent and novel nonpeptideantagonist of orexin-1 (OX(1)) receptor with Ki values of 27 nM and 57 nM in membrane formats and in whole cell, respectively. SB408124 was 50-fold selective over the human OX2receptor [1]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8688