Learn More
Cayman Chemical SB-431542, 301836-41-9, 1 mg
SDP
Supplier:  Cayman Chemical 130311
Encompass_Preferred
A potent and selective inhibitor of the TGF-β1 receptor ALK5 (IC50 = 94 nM), ALK4 (IC50 = 140 nM) and, less effectively, ALK7; suppresses renewal in embryonic and induced pluripotent stem cells and promotes their differentiation
Catalog No. NC0406544
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.