Learn More
Abcam SB431542 (DMSO solution), ALK inhibitor, 1MG-LQD
Supplier:  Abcam AB1465901MGLQD
Encompass_Preferred
MW 384.4 Da, Purity >99%. Potent and selective inhibitor of Activin Like Kinase. Inhibits ALK 4, 5 (IC₅₀ = 94 nM), and 7.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1903553