Learn More
Selleck Chemical LLC SB505124 10mg 694433-59-5
SDP
Supplier:  Selleck Chemical LLC S218610MG
Encompass_Preferred
SB505124 is a selective inhibitor of TGF?R for ALK4, ALK5 with IC50 of 129 nM and 47 nM in cell-free assays, respectively, also inhibits ALK7, but does not inhibit ALK1, 2, 3, or 6. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7991