Learn More
Selleck Chemical LLC SB590885 10mg 405554-55-4
SDP
Supplier:  Selleck Chemical LLC S222010MG
Encompass_Preferred
SB590885 is a potent B-Raf inhibitor with Ki of 0.16 nM in a cell-free assay, 11-fold greater selectivity for B-Raf over c-Raf, no inhibition to other human kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-838-6