Learn More
Medchemexpress LLC HY-102016 10mg Medchemexpress, SB756050 CAS:447410-57-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10201610MG
Encompass_Preferred
Medchemexpress, HY-102016 10mg SB756050 CAS:447410-57-3 SB756050 is a selective TGR5 agonist. SB756050 has the potential for type 2 diabetes treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0647